Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rptoros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045312.1 | 4 | 954 | |||
NR_045313.1 | 3 | 839 | ||||
GenBank mRNA | AK040159.1 | - | 4 | 946 | ||
AK086908.1 | - | 3 | 840 | |||
BY433329.1 | - | 1 | 61 |