Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610028E06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015560.1 | 6 | 822 | |||
GenBank mRNA | AK011585.1 | - | 5 | 712 | ||
AK031953.1 | - | 6 | 822 | |||
AK082619.1 | - | 6 | 747 | |||
AK083757.1 | - | 7 | 1235 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY754716.1 | - | 4 | 480 | Chr. 4 - 125890415 - 125893958 [-] on Build GRCm38 |