Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045366.1 | 4, 5 | 478 | |||
GenBank mRNA | BC048666.1 | - | 4, 5 | 478 |