Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040613.1 | 2 | 203 | |||
GenBank mRNA | AK015552.1 | - | 2 | 205 | ||
BY715100.1 | - | 2 | 205 |