Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frs3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045912.1 | 1 | 106 | |||
NR_045913.1 | 1 | 106 | ||||
GenBank mRNA | AK076701.1 | - | 1 | 107 |