Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A230056P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015495.2 | 5 | 637 | |||
GenBank mRNA | AK043630.1 | - | 5 | 638 | ||
AK140288.1 | - | 5 | 638 | |||
BC147226.1 | - | 3 | 276 | |||
BC147227.1 | - | 3 | 276 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727408.1 | - | 5 | 627 | Chr. 7 - 55962541 - 55976908 [+] on Build GRCm38 |