Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080699.1 | - | 347 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742117.1 | - | 2 | 343 | Chr. 1 - 19064924 - 19104837 [-] on Build GRCm38 |