Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI503316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040295.1 | - | 2 | 800 | ||
BB200448.2 | - | 1 | 370 | |||
BC033430.1 | - | 2 | 1093 |