Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9130221H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046001.1 | 1 | 1501 | |||
GenBank mRNA | AK020285.1 | - | 1 | 1206 | ||
BC022632.1 | - | 2 | 1426 | 4207058 |
Gene Symbol: Zfp526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK173308.1 | - | 2685 |