Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mir99ahg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015543.2 | 4 | 624 | |||
GenBank mRNA | AK012955.1 | - | 497 | |||
AK021336.1 | - | 668 | ||||
AK033222.1 | - | 229 | ||||
AK048031.1 | - | 679 | ||||
AK083771.1 | - | 4 | 625 | |||
AK144299.1 | - | 644 | ||||
AK148800.1 | - | 565 |