Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015480.1 | 1 | 105 | |||
GenBank mRNA | AK042966.1 | - | 1 | 105 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB640217.1 | - | 1 | 105 | Chr. 7 - 45366161 - 45367875 [+] on Build GRCm38 |