Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1500015L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045817.2 | 2 | 218 | |||
GenBank mRNA | AK005254.1 | - | 2 | 219 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_183277.1 | - | 219 | Chr. - - [-] on Build |