Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033481.1 | 2 | 280 | |||
GenBank mRNA | AK148387.1 | - | 2 | 281 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY741516.1 | - | 2 | 278 | Chr. X - 68669523 - 68678397 [-] on Build GRCm38 |