Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700086O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015475.1 | 1 | 415 | |||
NR_027903.1 | 1 | 415 | ||||
GenBank mRNA | AK007023.1 | - | 1 | 418 | ||
AK050707.1 | - | 1 | 390 | |||
AK078369.1 | - | 1 | 415 |
Gene Symbol: Ttc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC035338.1 | - | 356 |