Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep83os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015524.1 | 2 | 461 | |||
GenBank mRNA | AK015136.1 | - | 2 | 375 | ||
AK016525.1 | - | 2 | 420 | |||
AK085032.1 | - | 2 | 426 | |||
AK144701.1 | - | 2 | 423 | |||
AK154718.1 | - | 2 | 393 | |||
AK155868.1 | - | 2 | 393 | |||
BC067070.1 | - | 2 | 406 |