Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9930014A18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030696.1 | 3 | 3361 | |||
GenBank mRNA | AK035741.1 | - | 1 | 559 | ||
AK036813.1 | - | 2 | 1958 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177166.1 | - | 559 | Chr. - - [-] on Build |