Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4921504A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102341.1 | 4 | 1504 | |||
NR_102342.1 | 3 | 1416 | ||||
GenBank mRNA | AK046351.1 | - | 4 | 1504 |