Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810429I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015522.1 | 3 | 150 | |||
GenBank mRNA | AK013191.1 | - | 158 | |||
AK031301.1 | - | 136 | ||||
AK034854.1 | - | 1 | 1714 | |||
AK043092.1 | - | 3 | 232 | |||
AK043278.1 | - | 3 | 151 | |||
AK084238.1 | - | 1 | 1724 | |||
AK087718.1 | - | 2 | 145 |