Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045969.1 | 4 | 1883 | |||
NR_045970.1 | 4 | 1859 | ||||
NR_045971.1 | 4 | 1856 | ||||
NR_045972.1 | 3 | 1747 | ||||
GenBank mRNA | AK085722.1 | - | 1749 | |||
BC030866.1 | - | 1 | 985 | |||
BC040816.1 | - | 1 | 973 |