Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700025M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040687.1 | 5 | 523 | |||
NR_040688.1 | 4 | 431 | ||||
NR_040689.1 | 3 | 351 | ||||
GenBank mRNA | AK006359.1 | - | 3 | 354 | ||
BC048655.1 | - | 5 | 520 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA465171.1 | - | 523 | Chr. - - [-] on Build |