Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108067.1 | 5 | 1700 | |||
NR_108068.1 | 3 | 1439 | ||||
GenBank mRNA | AK123440.1 | - | 3 | 1439 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000626230.2 | - | 1439 | Chr. - - [-] on Build |