Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR325HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110400.2 | 1 | 99 | |||
NR_110401.2 | 1 | 99 | ||||
NR_110402.2 | 1 | 99 | ||||
NR_110403.2 | 1 | 99 | ||||
NR_110404.2 | 1 | 99 | ||||
NR_110405.2 | 1 | 99 | ||||
NR_110406.2 | 1 | 99 | ||||
GenBank mRNA | AK057746.1 | - | 1 | 218 | ||
AK311342.1 | - | 1 | 99 | |||
DA256943.1 | - | 1 | 69 | |||
DA300762.1 | - | 1 | 67 | |||
DA388267.1 | - | 1 | 35 | |||
DA389464.1 | - | 1 | 67 |