Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR325HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110400.2 | 4 | 1050 | |||
NR_110401.2 | 4 | 1034 | ||||
NR_110402.2 | 4 | 988 | ||||
NR_110403.2 | 4 | 966 | ||||
NR_110404.2 | 3 | 929 | ||||
NR_110405.2 | 3 | 872 | ||||
NR_110406.2 | 3 | 867 | ||||
GenBank mRNA | AK057746.1 | - | 2 | 490 | ||
AK311342.1 | - | 4 | 988 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000630388.2 | - | 988 | Chr. - - [-] on Build |