Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110533.1 | 6 | 1238 | |||
GenBank mRNA | X91348.1 | - | LIT1227 DGCR5 |
6 | 1209 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000440005.6 | - | 6 | 1211 | Chr. 22 - 18970525 - 19031242 [+] on Build GRCh38 |