Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKILA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131157.1 | 1 | 1157 | |||
GenBank mRNA | AK056098.1 | - | 1 | 1157 |
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL732105.1 | - | 2 | 132 |