Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNM3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038397.2 | 1 | 6219 | |||
NR_103486.1 | 4 | 2313 | ||||
GenBank mRNA | FJ623959.1 | - | 1 | 6219 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA439854.1 | - | 1 | 524 | Chr. 1 - 172138578 - 172139131 [-] on Build GRCh38 |