Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120632.1 | 4 | 1882 | |||
GenBank mRNA | AK055803.1 | - | 4 | 1883 |