Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYT1L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024468.1 | 2 | 1161 | |||
GenBank mRNA | AK057398.1 | - | 2 | 1081 | ||
AK126008.1 | - | 2 | 1149 |