Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110915.1 | 3 | 388 | |||
GenBank mRNA | BC039386.1 | - | 3 | 388 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI561273.1 | - | 3 | 392 | Chr. 16 - 14019087 - 14021067 [-] on Build GRCh38 |