Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026543.1 | 3 | 643 | |||
GenBank mRNA | BC062296.1 | - | 3 | 645 | ||
BC080530.1 | - | 3 | 642 | |||
BG751912.1 | - | 3 | 642 |