Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC8A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038441.1 | 5 | 307 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI837006.1 | - | 5 | 305 | Chr. 2 - 39917634 - 40254927 [+] on Build GRCh38 |