Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033965.1 | 2 | 437 | |||
GenBank mRNA | AK057627.1 | - | 2 | 218 | ||
AK309698.1 | - | 2 | 251 | |||
AK309874.1 | - | 2 | 247 | |||
BC031275.1 | - | 2 | 200 | |||
BC045736.1 | - | 2 | 281 | |||
BC106900.2 | - | 2 | 437 | |||
BC118565.1 | - | 2 | 430 |