Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033202.2 | 2 | 258 | |||
NR_033203.1 | 2 | 161 | ||||
NR_033205.1 | 2 | 254 | ||||
NR_110331.1 | 2 | 258 | ||||
GenBank mRNA | AA299267.1 | - | 2 | 198 | ||
BG196429.1 | - | 1 | 63 | |||
BG203124.1 | - | 2 | 289 | |||
BG203609.1 | - | 2 | 284 | |||
CB046509.1 | - | 2 | 170 | |||
CF127739.1 | - | 2 | 236 | |||
DA741273.1 | - | 2 | 263 |