Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110787.1 | 3 | 691 | |||
GenBank mRNA | BC042016.1 | - | 3 | 691 |