Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000701.1 | - | 1 | 158 | ||
AK222717.1 | - | 1 | 156 | |||
BC128165.1 | - | 1 | 57 | |||
BC128166.1 | - | 1 | 57 |