Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105371899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136416.1 | 1 | 201 | |||
NR_136417.1 | 1 | 201 | ||||
GenBank mRNA | BM700476.1 | - | 1 | 99 | ||
CD675441.1 | - | 1 | 42 | |||
CD675892.1 | - | 1 | 158 | |||
DB157978.1 | - | 1 | 201 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK123869.1 | - | 1 | 532 | Chr. 17 - 76966285 - 76969530 [-] on Build GRCh38 |