Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108043.1 | 1, 2 | 41 | |||
GenBank mRNA | BX089194.1 | - | 1, 2 | 41 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI792816.1 | - | 1, 2 | 42 | Chr. 10 - 34815767 - 34816386 [+] on Build GRCh38 |