Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125878.1 | 3 | 513 | |||
GenBank mRNA | BX092407.1 | - | 3 | 483 |