Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JARID2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120502.1 | 2 | 439 | |||
GenBank mRNA | BC033996.1 | - | 2 | 400 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY043093.1 | - | 2 | 413 | Chr. 6 - 15247832 - 15248669 [-] on Build GRCh38 |