Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023925.1 | 6, 7 | 690 | |||
NR_023926.1 | 5, 6 | 644 | ||||
NR_023927.1 | 3, 4 | 349 | ||||
NR_110327.1 | 4, 5 | 461 | ||||
GenBank mRNA | AF295725.1 | - | 6, 7 | 690 | ||
AF295726.1 | - | 5, 6 | 605 | |||
AF295727.1 | - | 4, 5 | 393 | |||
AF295728.1 | - | 5, 6 | 337 | |||
AF295729.1 | - | 3, 4 | 220 | |||
AF295730.1 | - | 3, 4 | 371 | |||
AK057505.1 | - | 4, 5 | 493 |