Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024127.1 | 1, 2 | 276 | |||
GenBank mRNA | AF277181.1 | - | 1, 2 | 64 | ||
AK092096.1 | - | 1 | 657 | |||
BC022205.1 | - | 1, 2 | 905 | |||
BC034611.1 | - | 1, 2 | 86 | |||
BC045792.1 | - | 1, 2 | 294 | |||
BC064589.1 | - | 1, 2 | 264 | |||
BC068048.1 | - | 1, 2 | 276 | |||
KJ782215.1 | - | 2, 3 | 1405 |