Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108062.1 | 3 | 550 | |||
NR_108063.1 | 3 | 550 | ||||
GenBank mRNA | BC017942.1 | - | 3 | 545 | ||
BC018008.1 | - | 3 | 545 | |||
BC020935.1 | - | 3 | 541 | |||
BC026300.1 | - | 2 | 346 | |||
CX163049.1 | - | 3 | 551 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX167424.1 | - | 3 | 499 | Chr. 13 - 54126963 - 54132820 [-] on Build GRCh38 |