Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105369340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135086.1 | 2 | 454 | |||
NR_135087.1 | 2 | 472 | ||||
GenBank mRNA | BG206029.1 | - | 1 | 254 |