Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC115110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037844.2 | 5 | 3830 | |||
GenBank mRNA | AK021767.1 | - | 1 | 840 | ||
AL359943.1 | - | 1 | 2881 | |||
BC101995.1 | - | 1 | 332 | |||
BC101996.1 | - | 1 | 332 | |||
BC101997.1 | - | 1 | 332 | |||
BC103697.1 | - | 1 | 332 | |||
BC114357.1 | - | 1 | 332 | |||
BC114358.2 | - | 1 | 332 | |||
CR749717.1 | - | 1 | 2881 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI045534.1 | - | 1 | 125 | Chr. 1 - 2550583 - 2551391 [-] on Build GRCh38 |