Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCIRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017089.1 | XP_016872578.1 | 20 | 3280 | ||
GenBank mRNA | AK057093.1 | - | 1 | 2053 |