Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135252.1 | 1 | 229 | |||
GenBank mRNA | BC040628.1 | - | 1 | 229 |