Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100129175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046086.1 | 3 | 1946 | |||
GenBank mRNA | AK092605.1 | - | 3 | 1946 |