Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105369486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135100.1 | 1 | 47 | |||
GenBank mRNA | DA016239.1 | - | 1 | 48 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA016285.1 | - | 1 | 31 | Chr. 11 - 110355130 - 110366485 [+] on Build GRCh38 |