Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125887.1 | 11, 12 | 2180 | |||
GenBank mRNA | BC040325.1 | - | 11, 12 | 2180 |