Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100128233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103769.1 | 2, 3 | 320 | |||
GenBank mRNA | BX161428.1 | - | 2, 3 | 320 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX355977.2 | - | 2, 3 | 322 | Chr. 14 - 65213643 - 65222347 [-] on Build GRCh38 |